8-Nitroquinoline

Supplier: Molekula

MOLEM38941435EA 0 GBP
MOLEM38941435
8-Nitroquinoline
8-Nitroquinoline
Formula: C₉H₆N₂O₂
MW: 174,16 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 89…91 °C
MDL Number: MFCD00006806
CAS Number: 607-35-2
EINECS: 210-135-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR