3-Quinolinecarboxylic acid ≥98%

Supplier: Thermo Scientific
Warning

L15133.03 L15133.06
L15133.03EA 37.9 GBP
L15133.03 L15133.06
3-Quinolinecarboxylic acid ≥98%
3-Quinolinecarboxylic acid

Purity: 98% CAS No.: 6480-68-8 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: ca 283° dec. Beilstein Reference: 126542

Formula: C₁₀H₇NO₂
MW: 173,17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: ∼ 283 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006770
CAS Number: 6480-68-8
EINECS: 229-337-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR