3-Bromoquinoline 98%
Supplier: Thermo Scientific
|
Warning
|
154270250
154271000
AC15427.1
AC15427.10
AC15427.50
ACRO154270250EA
134
GBP
ACRO154270250
ACRO154271000
MAYBAC15427.1
MAYBAC15427.10
MAYBAC15427.50
3-Bromoquinoline 98%
3-Bromoquinoline
Formula:
C₉H₆BrN MW: 208,06 g/mol Boiling Pt: 274 °C (1013 hPa) Melting Pt: 13…15 °C Density: 1,53 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006767 CAS Number: 5332-24-1 EINECS: 226-237-1 UN: 2810 |
Specification Test Results
Appearance | Clear colorless to yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6630 to 1.6660 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...