1,4-Diethylbenzene, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

32018-5ML
32018-5MLEA 216 GBP
32018-5ML
1,4-Diethylbenzene, Supelco®
1,4-Diethylbenzene
Formula: C₁₀H₁₄
MW: 134,22 g/mol
Boiling Pt: 183…184 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0,862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009264
CAS Number: 105-05-5
EINECS: 203-265-2
UN: 2049
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96afc1b50e846e1670ba1301ed7928f88450afa6&vid=6b8c33fb8d3e66ac232184e06700c40456db83a4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR