3-Aminobenzamidoxime 97%

Supplier: Thermo Fisher Scientific
Warning

H51773.03 H51773.06
H51773.03EA 0 GBP
H51773.03 H51773.06
3-Aminobenzamidoxime 97%
3-Aminobenzamidoxime

Purity: 97% CAS No.: 100524-07-0 Molecular Formula: C7H9N3O Formula Weight: 151.17 Melting Point: 101-105° MDL No.: MFCD08061150

Formula: C₇H₉N₃O
MW: 151.17 g/mol
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD08061150
CAS Number: 100524-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR