1,3-Diphenylurea ≥98%
Supplier: Thermo Scientific
Synonyms:
N,N’-Diphenylurea, N,N'-Diphenylurea
A18720.14
A18720.22
A18720.14EA
32
GBP
A18720.14
A18720.22
1,3-Diphenylurea ≥98%
1,3-Diphenylurea
Purity: 98%
CAS No.: 102-07-8
Molecular Formula: C6H5NHCONHC6H5
Formula Weight: 212.25
Melting Point: 240-244°
Boiling Point: 260° dec.
Merck Reference: 14,1785
MDL No.: MFCD00003017
Formula:
C₁₃H₁₂N₂O MW: 212,25 g/mol Boiling Pt: 260 °C (dec.) Melting Pt: 240…244 °C Density: 1,239 Storage Temperature: Ambient |
MDL Number:
MFCD00003017 CAS Number: 102-07-8 EINECS: 203-003-7 Merck Index: 14,1785 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...