4-Bromophenethylamine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-(4-Bromophenyl)ethylamine
183090010
183090050
ACRO183090010EA
75.8
GBP
ACRO183090010
ACRO183090050
4-Bromophenethylamine 98%
4-Bromophenethylamine
Formula:
C₈H₁₀BrN MW: 200.08 g/mol Boiling Pt: 63…72 °C (.2 mmHg) Density: 1.29 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008189 CAS Number: 73918-56-6 EINECS: 277-636-2 UN: 2735 ADR: 8 III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5730 to 1.5750 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...