4-Chlorophenethylamine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-(4-Chlorophenyl)ethylamine

109880250 109880050
ACRO109880250EA 121 GBP
ACRO109880250 ACRO109880050
4-Chlorophenethylamine 98%
4-Chlorophenethylamine
Formula: C₈H₁₀ClN
MW: 155,63 g/mol
Boiling Pt: 242…245 °C (760 mmHg)
Density: 1,112 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5474 to 1.5494 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR