2-Isopropylphenol 97%
Supplier: Thermo Scientific
|
Danger
|
156680050
156685000
ACRO156680050EA
37.4
GBP
ACRO156680050
ACRO156685000
2-Isopropylphenol 97%
2-Isopropylphenol
Formula:
C₉H₁₂O MW: 136,19 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: 15…16 °C Density: 1,01 g/cm³ Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002224 CAS Number: 88-69-7 EINECS: 201-852-8 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5250 to 1.5290 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...