2-Ethylaniline 98%

Supplier: Thermo Scientific
Danger

118050010 118050050 118052500
ACRO118050010EA 471 GBP
ACRO118050010 ACRO118050050 ACRO118052500
2-Ethylaniline 98%
2-Ethylaniline
Formula: C₈H₁₁N
MW: 121,18 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0,98 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS Number: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥97.5 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR