3-Methyl-4-nitrobenzoic acid ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Methyl-4-nitrobenzoic acid, 4-Nitro-m-toluic acid

A15523.22 A15523.36
A15523.22EA 52.7 GBP
A15523.22 A15523.36
3-Methyl-4-nitrobenzoic acid ≥99%
4-Nitro-m-toluic acid

Purity: 99% CAS No.: 3113-71-1 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 215-220° Beilstein Reference: 1959154

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 356
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007168
CAS Number: 3113-71-1
EINECS: 221-479-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR