4-Hydroxybenzylacetone ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone
A14064.14
A14064.22
A14064.30
A14064.14EA
45.6
GBP
A14064.14
A14064.22
A14064.30
4-Hydroxybenzylacetone ≥98%
4-Hydroxybenzylacetone
Purity: 98%
CAS No.: 5471-51-2
Molecular Formula: C10H12O2
Formula Weight: 164.20
Melting Point: 80-84°
Beilstein Reference: 776080
Formula:
C₁₀H₁₂O₂ MW: 164,2 g/mol Boiling Pt: 292 °C (1013 hPa) Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002394 CAS Number: 5471-51-2 EINECS: 226-806-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...