4-Tolualdehyde 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde

139001000 139005000 139000025
ACRO139001000EA 51.2 GBP
ACRO139001000 ACRO139005000 ACRO139000025
4-Tolualdehyde 97%
4-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 204…205 °C (760 mmHg)
Melting Pt: –6 °C
Density: 1,015 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5440 to 1.5480 (20°C, 589 nm)
GC ≥96.0 %
Specific gravity (20°C) 1.010 to 1.025


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR