2-Isopropoxyphenylboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Isopropoxybenzeneboronic acid, (2-Isopropoxyphenyl)boronic acid

H53168.03 H53168.06
H53168.03EA 35.6 GBP
H53168.03 H53168.06
2-Isopropoxyphenylboronic acid 95%
2-Isopropoxyphenylboronic acid

Purity: 95% CAS No.: 138008-97-6 Molecular Formula: C9H13BO3 Formula Weight: 180.01 Flash Point: 110°(230°F) Refractive Index: 1.5090 Melting Point: 30-35° Boiling Point: 199° MDL No.: MFCD03427050

Formula: C₉H₁₃BO₃
MW: 180,01 g/mol
Boiling Pt: 199 °C
Melting Pt: 30…35 °C
Density: 1,075
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03427050
CAS Number: 138008-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR