1,4,7,10-Tetraazacyclododecane 97%
Supplier: Thermo Scientific
|
Danger
|
294322500
294320010
294320500
ACRO294322500EA
275
GBP
ACRO294322500
ACRO294320010
ACRO294320500
1,4,7,10-Tetraazacyclododecane 97%
1,4,7,10-Tetraazacyclododecane (Cyclen, Kryptofix® 11 aza)
Formula:
C₈H₂₀N₄ Melting Pt: 108…113 °C Storage Temperature: Freezer |
MDL Number:
MFCD00066281 CAS Number: 294-90-6 EINECS: 425-450-9 UN: 1759 ADR: 8,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 108 to 113 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...