Diphenyl sulphoxide 97%

Supplier: Thermo Scientific

Synonyms: Phenyl sulphoxide, sulphinyldibenzene

130970250 130971000
ACRO130970250EA 114 GBP
ACRO130970250 ACRO130971000
Diphenyl sulphoxide 97%
Diphenyl sulphoxide
Formula: C₁₂H₁₀OS
MW: 202,28 g/mol
Boiling Pt: 206…208 °C (13 mmHg)
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002085
CAS Number: 945-51-7
EINECS: 213-415-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 66 to 71 °C
Water ≤0.5 % (K.F.)
GC ≥96.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR