4'-(Trifluoromethyl)biphenyl-2-carboxylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
B25099.03
B25099.06
B25099.03EA
36.2
GBP
B25099.03
B25099.06
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid ≥98%
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
Purity: 98%
CAS No.: 84392-17-6
Molecular Formula: C14H9F3O2
Formula Weight: 266.22
Melting Point: 167-170°
Beilstein Reference: 7577560
Formula:
C₁₄H₉F₃O₂ MW: 266,22 g/mol Melting Pt: 167…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075353 CAS Number: 84392-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...