Diethyl ethoxymethylenemalonate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Diethyl 2-(ethoxymethylene)malonate

A13776.0B A13776.18 A13776.22 A13776.30 A13776.36
A13776.0BEA 177 GBP
A13776.0B A13776.18 A13776.22 A13776.30 A13776.36
Diethyl ethoxymethylenemalonate ≥98%
Diethyl ethoxymethylenemalonate

Purity: 98+% CAS No.: 87-13-8 Molecular Formula: C10H16O5 Formula Weight: 216.23 Flash Point: 144°(291°F) Refractive Index: 1.4620 Odour: Ester-like Melting Point: -33° Boiling Point: 136-139°/10mm Beilstein Reference: 880058 MDL No.: MFCD00009148

Formula: C₁₀H₁₆O₅
Boiling Pt: 136…139 °C (10 mmHg)
Melting Pt: –33 °C
Density: 1,080
Flash Pt: 144 °C (291 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR