Dimethyl fluoromalonate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Dimethyl 2-fluoromalonate
L16751.03
L16751.06
L16751.03EA
61.5
GBP
L16751.03
L16751.06
Dimethyl fluoromalonate ≥97%
Dimethyl fluoromalonate
Purity: 97%
CAS No.: 344-14-9
Molecular Formula: C5H7FO4
Formula Weight: 150.11
Refractive Index: 1.4032
Boiling Point: 111-112°/45mm
UN No.: 3261
MDL No.: MFCD00054680
Formula:
C₅H₇FO₄ MW: 150,11 g/mol Boiling Pt: 111…112 °C (45 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00054680 CAS Number: 344-14-9 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...