3-Nitroanthranilic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-3-nitrobenzoic acid
H26001.03
H26001.09
H26001.03EA
24.7
GBP
H26001.03
H26001.09
3-Nitroanthranilic acid 97%
3-Nitroanthranilic acid
Purity: 97%
CAS No.: 606-18-8
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Melting Point: 210-212°
MDL No.: MFCD00024261
Formula:
C₇H₆N₂O₄ MW: 182,14 g/mol Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024261 CAS Number: 606-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...