1,2,3-Trimethoxybenzene 98%

Supplier: Thermo Scientific

160210250 160211000
ACRO160210250EA 65.6 GBP
ACRO160210250 ACRO160211000
1,2,3-Trimethoxybenzene 98%
1,2,3-Trimethoxybenzene
Formula: C₉H₁₂O₃
MW: 168,19 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 1,112 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008358
CAS Number: 634-36-6
EINECS: 211-207-2

Order Now

Specification Test Results

Appearance White to beige to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR