Dibutyl sulphoxide ≥97%

Supplier: Thermo Scientific

Synonyms: n-Butyl sulphoxide, Di-n-butyl sulphoxide, Butyl sulphoxide

B23672.06 B23672.14
B23672.06EA 32.2 GBP
B23672.06 B23672.14
Dibutyl sulphoxide ≥97%
Dibutyl sulphoxide

Purity: 97% CAS No.: 2168-93-6 Molecular Formula: C8H18OS Formula Weight: 162.30 Flash Point: 121°(249°F) Refractive Index: 1.4670 Melting Point: 31-34° Boiling Point: 250° Beilstein Reference: 1747425 MDL No.: MFCD00002093

Formula: C₈H₁₈OS
MW: 162,3 g/mol
Boiling Pt: 250 °C
Melting Pt: 31…34 °C
Density: 0,832
Flash Pt: 121 °C (249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002093
CAS Number: 2168-93-6
EINECS: 218-511-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR