Methylmalonic acid 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Methylmalonic acid
127090050
127090250
127091000
ACRO127090050EA
76
GBP
ACRO127090050
ACRO127090250
ACRO127091000
Methylmalonic acid 96%
Methylmalonic acid
Formula:
C₄H₆O₄ MW: 118,09 g/mol Melting Pt: 129…136 °C Density: 1,45 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00002656 CAS Number: 516-05-2 EINECS: 208-219-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 129 to 136 °C |
Water | ≤0.5 % (K.F.) |
Titration with NaOH | ≥95 % |
Color scale | ≤100 APHA |
Acidity | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...