Manganese ≥99.9% (metals basis), nanopowder APS 30-50 nm
Supplier: Thermo Scientific
|
Warning
|
45508.06
45508.14
45508.06EA
258
GBP
45508.06
45508.14
Manganese ≥99.9% (metals basis), nanopowder APS 30-50 nm
Manganese
Purity: 99.9%
CAS No.: 7439-96-5
Molecular Formula: Mn(core)/Mn3O4(shell)
Physical Form: Mn3O4 layer 5-12nm
UN No.: 3089
MDL No.: MFCD00011111
Formula:
Mn MW: 54,94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...