2-Ethoxyphenol 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol

370030050 370035000
ACRO370030050EA 45 GBP
ACRO370030050 ACRO370035000
2-Ethoxyphenol 98%
2-Ethoxyphenol
Formula: C₈H₁₀O₂
Boiling Pt: 216…217 °C
Melting Pt: 20…25 °C
Density: 1,09 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR