2,3,4-Trifluoroaniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10254.06 A10254.14
A10254.06EA 0 GBP
A10254.06 A10254.14
2,3,4-Trifluoroaniline ≥98%
2,3,4-Trifluoroaniline

Purity: 98% CAS No.: 3862-73-5 Molecular Formula: C6H4F3N Formula Weight: 147.10 Flash Point: 68°(154°F) Refractive Index: 1.4870 Boiling Point: 90-92°/48mm Beilstein Reference: 3245609 UN No.: 3082 MDL No.: MFCD00011737

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 90…92 °C (48 mmHg)
Melting Pt: 14…15 °C
Density: 1.393
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR