1,2,4,5-Tetrafluorobenzene

Supplier: Molekula

41236301-25G 41236301-100G
MOLE41236301-25GEA 68.9 GBP
MOLE41236301-25G MOLE41236301-100G
1,2,4,5-Tetrafluorobenzene
1,2,4,5-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150,08 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00000307
CAS Number: 327-54-8
EINECS: 206-319-3
UN: 2387
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR