3,5-Dichlorophenol ≥98%
Supplier: Thermo Scientific
|
Warning
|
A15328.06
A15328.14
A15328.18
A15328.06EA
48.8
GBP
A15328.06
A15328.14
A15328.18
3,5-Dichlorophenol ≥98%
3,5-Dichlorophenol
Purity: 98%
CAS No.: 591-35-5
Molecular Formula: C6H4Cl2O
Formula Weight: 163.00
Odour: Phenol-like
Melting Point: 63-68°
Boiling Point: 233°
Beilstein Reference: 1906458
UN No.: 2020
MDL No.: MFCD00002259
Formula:
C₆H₄Cl₂O MW: 163 g/mol Boiling Pt: 233 °C Melting Pt: 63…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002259 CAS Number: 591-35-5 EINECS: 209-714-9 UN: 2020 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...