2-Chloroanisole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Chloro-2-methoxybenzene

L06952.18 L06952.30
L06952.18EA 63.2 GBP
L06952.18 L06952.30
2-Chloroanisole ≥98%
2-Chloroanisole
Formula: C₇H₇ClO
MW: 142,58 g/mol
Boiling Pt: 194…196 °C
Melting Pt: –27 °C
Density: 1,123
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000557
CAS Number: 766-51-8
EINECS: 212-167-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR