1-Chloro-2-fluorobenzene ≥98%
Supplier: Thermo Scientific
|
Warning
|
B21209.14
B21209.36
B21209.14EA
135
GBP
B21209.14
B21209.36
1-Chloro-2-fluorobenzene ≥98%
1-Chloro-2-fluorobenzene
Purity: 98+%
CAS No.: 348-51-6
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 31°(87°F)
Refractive Index: 1.5010
Melting Point: -43°
Boiling Point: 137-138°
Beilstein Reference: 1855301
UN No.: 1993
MDL No.: MFCD00000533
Formula:
C₆H₄ClF MW: 130,55 g/mol Boiling Pt: 137…138 °C Melting Pt: –43 °C Density: 1,242 Flash Pt: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000533 CAS Number: 348-51-6 EINECS: 206-476-8 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...