2-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.20276.0250
8.20276.1000
8.20276.9025
8.20276.0250EA
40.5
GBP
8.20276.0250
8.20276.1000
8.20276.9025
2-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
2-Chlorobenzaldehyde
Formula:
C₇H₅ClO MW: 140,57 g/mol Boiling Pt: 211,9 °C (1013 hPa) Melting Pt: 10 °C Density: 1,248 g/cm³ (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003304 CAS Number: 89-98-5 EINECS: 201-956-3 UN: 3265 ADR: 8,III |
Specification Test Results
Solubility | 1.12 g/l (20 °C) |
Melting Point | 10 - 11.5 °C |
Molar Mass | 140.56 g/mol |
Boiling Point | 208 °C |
Vapor Pressure | 1.7 hPa (50 °C) |
Flash Point | 87 °C |
Refractive Index | 1.5658 (20 °C, 589 nm) |
Density | 1.25 g/cm3 (20 °C) |
pH | 2.9 (H2O) (saturated aqueous solution) |
Ignition Point | 397 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...