2-Chlorobenzaldehyde ≥97%
Supplier: Thermo Scientific
|
Danger
|
A13488.30
A13488.09
A13488.0B
A13488.30EA
25.5
GBP
A13488.30
A13488.09
A13488.0B
2-Chlorobenzaldehyde ≥97%
2-Chlorobenzaldehyde
Purity: 97%
CAS No.: 89-98-5
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 87°(188°F)
Refractive Index: 1.5675
Odour: Pungent
pH: 2.9
Melting Point: 10-12°
Boiling Point: 212°
Beilstein Reference: 385877
UN No.: 3265
MDL No.: MFCD00003304
Formula:
C₇H₅ClO MW: 140,57 g/mol Boiling Pt: 212 °C Melting Pt: 10…12 °C Density: 1,249 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003304 CAS Number: 89-98-5 EINECS: 201-956-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...