2,4-Dihydroxybenzoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
beta-Resorcylic acid
A13545.0E
A13545.22
A13545.36
A13545.0EEA
251
GBP
A13545.0E
A13545.22
A13545.36
2,4-Dihydroxybenzoic acid ≥97%
2,4-Dihydroxybenzoic acid
Purity: 97%
CAS No.: 89-86-1
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 226° dec.
Beilstein Reference: 1946213
Merck Reference: 14,8157
MDL No.: MFCD00002451
Formula:
C₇H₆O₄ MW: 154,12 g/mol Boiling Pt: 390 °C (1013 hPa) Melting Pt: ∼226 °C (dec.) Density: 1,54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002451 CAS Number: 89-86-1 EINECS: 201-946-9 Merck Index: 14,8157 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...