4-Ethoxybenzoic acid ≥98%

Supplier: Thermo Scientific
Warning

A12309.14 A12309.22 A12309.36
A12309.14EA 31.5 GBP
A12309.14 A12309.22 A12309.36
4-Ethoxybenzoic acid ≥98%
4-Ethoxybenzoic acid

Purity: 98+% CAS No.: 619-86-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 197-199° Beilstein Reference: 2207349

Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002545
CAS Number: 619-86-3
EINECS: 210-616-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR