4-(Dimethylamino)benzoic acid ≥98%
Supplier: Thermo Scientific
Synonyms:
4-Dimethylaminobenzoic acid
A14338.22
A14338.36
A14338.22EA
0
GBP
A14338.22
A14338.36
4-(Dimethylamino)benzoic acid ≥98%
4-(Dimethylamino)benzoic acid
Purity: 98+%
CAS No.: 619-84-1
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 170°(338°F)
Odour: Odourless
Beilstein Reference: 386867
Merck Reference: 14,3232
MDL No.: MFCD00002537
Formula:
C₉H₁₁NO₂ MW: 165,19 g/mol Boiling Pt: 297 °C (1013 hPa) Melting Pt: ∼240 °C (dec.) Density: 1,29 g/cm³ (20 °C) Flash Pt: 170 °C (338 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002537 CAS Number: 619-84-1 EINECS: 210-615-8 Merck Index: 14,3232 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...