4-(4-Nitrobenzyl)pyridine ≥98%
Supplier: Thermo Scientific
Synonyms:
4-(p-Nitrobenzyl)Pyridine
A13564.06
A13564.14
A13564.22
A13564.06EA
35.4
GBP
A13564.06
A13564.14
A13564.22
4-(4-Nitrobenzyl)pyridine ≥98%
4-(4-Nitrobenzyl)pyridine
Purity: 98+%
CAS No.: 1083-48-3
Molecular Formula: C12H10N2O2
Formula Weight: 214.22
Melting Point: 69-72°
Beilstein Reference: 170877
Formula:
C₁₂H₁₀N₂O₂ MW: 214,22 g/mol Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006445 CAS Number: 1083-48-3 EINECS: 214-108-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...