1-Bromo-3,4,5-trifluorobenzene ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Bromo-1,2,3-trifluorobenzene
B20607.03
B20607.06
B20607.03EA
48.9
GBP
B20607.03
B20607.06
1-Bromo-3,4,5-trifluorobenzene ≥98%
1-Bromo-3,4,5-trifluorobenzene
Purity: 98%
CAS No.: 138526-69-9
Molecular Formula: C6H2BrF3
Formula Weight: 210.98
Flash Point: 45°(113°F)
Refractive Index: 1.4820
Melting Point:
Formula:
C₆H₂BrF₃ MW: 210,98 g/mol Boiling Pt: 140 °C Melting Pt: < –20 °C Density: 1,767 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070738 CAS Number: 138526-69-9 EINECS: 418-480-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...