4-Bromobenzonitrile 98%

Supplier: Thermo Scientific
Warning

Synonyms: p-Bromobenzonitrile

106710100 106710500
ACRO106710100EA 59 GBP
ACRO106710100 ACRO106710500
4-Bromobenzonitrile 98%
4-Bromobenzonitrile
Formula: C₇H₄BrN
MW: 182,02 g/mol
Boiling Pt: 235…237 °C (760 mmHg)
Melting Pt: 111…114 °C
Density: 1,562 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00001811
CAS Number: 623-00-7
EINECS: 210-764-9
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or flakes
Infrared spectrum Conforms
Melting point 111 to 114 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR