3-Bromobenzonitrile 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
m-Bromobenzonitrile
106700100
106700025
106701000
ACRO106700100EA
86
GBP
ACRO106700100
ACRO106700025
ACRO106701000
3-Bromobenzonitrile 99%
3-Bromobenzonitrile
Formula:
C₇H₄BrN MW: 182,02 g/mol Boiling Pt: 225 °C (760 mmHg) Melting Pt: 37…41 °C Density: 1,562 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001796 CAS Number: 6952-59-6 EINECS: 230-127-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow to beige Crystalline powder or crystals or crystalline mass |
Infrared spectrum | Conforms |
Melting point | 37 to 41 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...