Hexabromobenzene ≥97%

Supplier: Thermo Fisher Scientific
Warning

B23295.14 B23295.22
B23295.14EA 29.1 GBP
B23295.14 B23295.22
Hexabromobenzene ≥97%
Hexabromobenzene

Purity: 97% CAS No.: 87-82-1 Molecular Formula: C6Br6 Formula Weight: 551.52 Odour: Odourless Melting Point: 300° Beilstein Reference: 1912586 MDL No.: MFCD00000058

Formula: C₆Br₆
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: >300 °C
Density: 3,543 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000058
CAS Number: 87-82-1
EINECS: 201-773-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR