Bromobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 108-86-1
Molecular Formula: C6H5Br
Formula Weight: 157.02
Flash Point: 51°(123°F)
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.5590
Odour: Aromatic
Melting Point: -31°
Boiling Point: 155-156°
Beilstein Reference: 1236661
Merck Reference: 14,1406
UN No.: 2514
MDL No.: MFCD00000055
Formula:
C₆H₅Br Boiling Pt: 155…156 °C Melting Pt: –31 °C Density: 1,492 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000055 CAS Number: 108-86-1 EINECS: 203-623-8 UN: 2514 ADR: 3,III Merck Index: 14,1406 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...