1-Dodecyne 97

Supplier: Thermo Fisher Scientific
Warning

L02977.06 L02977.14
L02977.06EA 48.8 GBP
L02977.06 L02977.14
1-Dodecyne 97
1-Dodecyne

Purity: 97% CAS No.: 765-03-7 Molecular Formula: CH3(CH2)9C≡CH Formula Weight: 166.31 Flash Point: 79°(174°F) Refractive Index: 1.4340 Melting Point: -19° Boiling Point: 214-216° Beilstein Reference: 1741320 MDL No.: MFCD00008960

Formula: C₁₂H₂₂
MW: 166.31 g/mol
Boiling Pt: 214…216 °C
Melting Pt: –19 °C
Density: 0.779
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008960
CAS Number: 765-03-7
EINECS: 212-134-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR