trans-5-Decene ≥97%

Supplier: Thermo Fisher Scientific
Warning

L10689.06 L10689.03
L10689.06EA 111 GBP
L10689.06 L10689.03
trans-5-Decene ≥97%
trans-5-Decene

Purity: 97% CAS No.: 7433-56-9 Molecular Formula: C10H20 Formula Weight: 140.27 Flash Point: 46°(114°F) Refractive Index: 1.4240 Boiling Point: 171-172° Beilstein Reference: 1719487 UN No.: 3295 MDL No.: MFCD00009508

Formula: C₁₀H₂₀
Boiling Pt: 171…172 °C
Melting Pt: –73 °C
Density: 0,740
Flash Pt: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009508
CAS Number: 7433-56-9
EINECS: 231-080-7
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR