1-Heptene ≥98%

Supplier: Thermo Scientific
Danger

L11487.AC L11487.AE
L11487.ACEA 61.6 GBP
L11487.AC L11487.AE
1-Heptene ≥98%
1-Heptene

Purity: 98+% CAS No.: 592-76-7 Molecular Formula: C7H14 Formula Weight: 98.19 Flash Point: -8°(17°F) Refractive Index: 1.400 Melting Point: ca -119° Boiling Point: 93-94° Beilstein Reference: 1098332 UN No.: 2278 MDL No.: MFCD00009531

Formula: C₇H₁₄
MW: 98,19 g/mol
Boiling Pt: 93…94 °C
Melting Pt: ∼–119 °C
Density: 0,697
Flash Pt: –8 °C (17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009531
CAS Number: 592-76-7
EINECS: 209-767-8
UN: 2278
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR