2,3-Dimethylbutane 98+%
Supplier: Thermo Fisher Scientific
|
Danger
|
116100050
116101000
116100010
ACRO116100050EA
40.7
GBP
ACRO116100050
ACRO116101000
ACRO116100010
2,3-Dimethylbutane 98+%
2,3-Dimethylbutane
Formula:
C₆H₁₄ Boiling Pt: 58 °C (760 mmHg) Melting Pt: –129 °C Density: 0,662 g/cm³ Flash Pt: –29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008925 CAS Number: 79-29-8 EINECS: 201-193-6 UN: 2457 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3740 to 1.3760 (20°C, 589 nm) |
GC | ≥98.0 % |
UV | at 260 nm A: ≤0.01 |
Titration after Ion exchange | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...