2,2,4-Trimethylpentane 99+% ACS

Supplier: Thermo Scientific
Danger

424560010 424565000 424560025
ACRO424560010EA 109 GBP
ACRO424560010 ACRO424565000 ACRO424560025
2,2,4-Trimethylpentane 99+% ACS
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 98…99 °C (760 mmHg)
Melting Pt: –107 °C
Density: 0,69 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=559f761f651e5fcdb0cf5dd303cd79e53daf2366&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR