n-Tridecane 99+%

Supplier: Thermo Scientific
Danger

Synonyms: Tridecane

139510250 139511000 139515000
ACRO139510250EA 75.3 GBP
ACRO139510250 ACRO139511000 ACRO139515000
n-Tridecane 99+%
n-Tridecane
Formula: H₃C(CH₂)₁₁CH₃
MW: 184,37 g/mol
Boiling Pt: 234 °C (760 mmHg)
Melting Pt: 5…4 °C
Density: 0,75 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008979
CAS Number: 629-50-5
EINECS: 211-093-4
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR