3-Chloro-1-phenyl-1-propyne ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(3-Chloroprop-1-yn-1-yl)benzene
H53510.14
H53510.03
H53510.06
H53510.14EA
369
GBP
H53510.14
H53510.03
H53510.06
3-Chloro-1-phenyl-1-propyne ≥97%
3-Chloro-1-phenyl-1-propyne
Purity: 97%
CAS No.: 3355-31-5
Molecular Formula: C9H7Cl
Formula Weight: 150.61
Flash Point: 104°(219°F)
Refractive Index: 1.585
MDL No.: MFCD06411085
Formula:
C₉H₇Cl MW: 150,61 g/mol Density: 1,095 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06411085 CAS Number: 3355-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...