1,13-Tetradecadiene ≥90%

Supplier: Thermo Scientific
Warning

Synonyms: Tetradeca-1,13-diene

H53479.03 H53479.06 H53479.14
H53479.03EA 59.7 GBP
H53479.03 H53479.06 H53479.14
1,13-Tetradecadiene ≥90%
1,13-Tetradecadiene

Purity: 90+% CAS No.: 21964-49-8 Molecular Formula: C14H26 Formula Weight: 194.36 Flash Point: 103°(217°F) Refractive Index: 1.443 Boiling Point: 131°/17mm Beilstein Reference: 1741459 MDL No.: MFCD00014941

Formula: C₁₄H₂₆
MW: 194,36 g/mol
Boiling Pt: 131 °C (17 mmHg)
Density: 0,849
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014941
CAS Number: 21964-49-8
EINECS: 244-687-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR