Nonylamine 97%

Supplier: Thermo Fisher Scientific
Danger

A18209.14 A18209.22 A18209.36
A18209.14EA 98.9 GBP
A18209.14 A18209.22 A18209.36
Nonylamine 97%
Nonylamine

Purity: 97+% CAS No.: 112-20-9 Molecular Formula: CH3(CH2)8NH2 Formula Weight: 143.27 Flash Point: 62°(143°F) Refractive Index: 1.4330 Odour: Amine-like Melting Point: -1° Boiling Point: 200-202° Beilstein Reference: 1733633 UN No.: 2735 MDL No.: MFCD00008249

Formula: H₃C(CH₂)₇CH₂NH₂
Boiling Pt: 200…202 °C
Melting Pt: –1 °C
Density: 0,784
Flash Pt: 62 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008249
CAS Number: 112-20-9
EINECS: 203-945-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR